Anti-MADH4 Goat Polyclonal Antibody

Supplier: GeneTex
GTX21341
89361-010EA 634.7 CAD
89361-010
Anti-MADH4 Goat Polyclonal Antibody
Smad4 is a common mediator of signal transduction by the TGF-beta (transforming growth factor) superfamily; it may act as a tumor suppressor.

For WB: Use at a concentration of 0.5-2?g/ml. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: MADH4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR