Anti-MUC1 Mouse Monoclonal Antibody [clone: M4H2]

Supplier: Genetex
GTX10120
89350-646EA 1147.57 CAD
89350-646
Anti-MUC1 Mouse Monoclonal Antibody [clone: M4H2]
Antibodies
Mouse monoclonal [M4H2] to MUC1

ELISA: Use at an assay dependent dilution. Can be used in ELISA pairs with GTX28322, 10117, 10123 and 10115. IF: Use at an assay dependent dilution. IHC: Use at an assay dependent dilution. Can be used for immunohistochemical staining of sections of breast, lung?and ovarian tumor tissues. Compatible with commonly used histological fixatives. IHC-P: Use at an assay dependent dilution. IHC-Fr: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: M4H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR