Anti-KRAS Mouse Monoclonal Antibody [clone: 4F3]

Supplier: GeneTex
GTX91809
89368-100EA 671.34 CAD
89368-100
Anti-KRAS Mouse Monoclonal Antibody [clone: 4F3]
Antibodies
Mouse monoclonal antibody [4F3] to K-Ras

For ELISA (tested against immunogen): Use at an assay dependent dilution. Not tested in other applications. For WB (tested against immunogen): Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: KRAS
Clonality: Monoclonal
Clone: 4F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR