Anti-MITF Chicken Polyclonal Antibody

GTX82132
89368-052EA 658.2 CAD
89368-052
Anti-MITF Chicken Polyclonal Antibody
Chicken polyclonal to Microphthalmia-associated transcription factor

WB (only tested on fusion protein): Use at a dilution of 1:2000. Colorimetric method for signal development.

Type: Primary
Antigen: MITF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR