Anti-IgG2a Rat Monoclonal Antibody

Supplier: GeneTex
GTX11571
89358-394EA 463.03 CAD
89358-394
Anti-IgG2a Rat Monoclonal Antibody
Antibodies
This secondary antibody specifically recognizes Mouse IgG2a. It does not react with any other isotypes.

Type: Secondary
Antigen: IgG2a
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy chain
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR