Anti-NGF Receptor Mouse Monoclonal Antibody [clone: NGFR5]

Supplier: GeneTex
GTX23125
89351-884EA 599.75 CAD
89351-884
Anti-NGF Receptor Mouse Monoclonal Antibody [clone: NGFR5]
Antibodies
NGF is important for the development, differentiation, and survival of variety of neuronal and non-neuronal cells. Its action is mediated by binding two distinct receptors, the high affinity p140 and low affinity Nerve Growth Factor (NGF) Receptor (p75 NGFR) or Neurotrophin Receptor (p75 NTR).

Type: Primary
Antigen: NGF Receptor
Clonality: Monoclonal
Clone: [NGFR5]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Baboon
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR