Anti-ORP1 Goat Polyclonal Antibody

Supplier: GeneTex
GTX10151
89349-088EA 918.77 CAD
89349-088
Anti-ORP1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to ORP1

Approx 48kDa band observed in Human Skeletal Muscle lysates (predicted MW of 50kDa according to NP_060500). Recommended for use at 0.5-2µg/ml. This antibody is expected to recognise both reported human isoforms of this protein.

Type: Primary
Antigen: ORP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR