Anti-HIP1 Mouse Monoclonal Antibody [clone: 1B11]

Supplier: GeneTex
GTX25403
89361-624EA 888.67 CAD
89361-624
Anti-HIP1 Mouse Monoclonal Antibody [clone: 1B11]
Antibodies
Mouse monoclonal [1B11] to Huntingtin Interacting Protein HIP1

For WB: Use at a dilution of 1:10,000. By Western blot, this antibody recognizes a band at ~115 kDa, representing HIP1 protein. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: HIP1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR