Anti-HIP1 Mouse Monoclonal Antibody [clone: 4B10]

Supplier: GeneTex

GTX25402
89361-622EA 762.95 CAD
89361-622
Anti-HIP1 Mouse Monoclonal Antibody [clone: 4B10]
Antibodies
Mouse monoclonal [4B10] to Huntingtin Interacting Protein HIP1

WB: Use at a dilution of 1/10000. Detects a band of approximately 120 kDa (predicted molecular weight: 115 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: HIP1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR