Anti-IgD Rabbit Polyclonal Antibody

Supplier: Genetex
GTX17184
89351-352EA 794.44 CAD
89351-352
Anti-IgD Rabbit Polyclonal Antibody
Antibodies
Little is known about the normal function of IgD, and few clinical signs or symptoms are associated with its absence. Individuals with low or absent levels of IgD do not appear unusually predisposed to infections. In mice, IgD may substitute for some functions of IgM when IgM is absent.

IHC-P: 1/40 - 1/100. Perform enzymatic antigen retrieval before commencing with IHC staining protocol. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IgD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR