Anti-IgE Mouse Monoclonal Antibody [clone: 4F4]

Supplier: GeneTex
GTX27294
89352-252EA 828.36 CAD
89352-252
Anti-IgE Mouse Monoclonal Antibody [clone: 4F4]
Antibodies
Mouse Monoclonal antibody [4F4] to Human IgE

For ELISA: Use at an assay dependent dilution. GTX27294 can be used for the detection of total IgE level and IgE antibodies in ELISA in combination with GTX27295, coating GTX27295 - conjugate GTX27294. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: IgE
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR