Anti-IGFBP2 Mouse Monoclonal Antibody [clone: 1.A.16]

Supplier: GeneTex
GTX24243
89361-434EA 754.55 CAD
89361-434
Anti-IGFBP2 Mouse Monoclonal Antibody [clone: 1.A.16]
Antibodies
Mouse monoclonal [1.A.16] to IGFBP2

ELISA: Use at an assay dependant dilution. WB: Use at an assay dependant dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IGFBP2
Clonality: Monoclonal
Clone: 1.A.16
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR