Anti-IGG Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX20938
89351-594EA 673.23 CAD
89351-594
Anti-IGG Rabbit Polyclonal Antibody
Antibodies
This antibody has been pretitered and quality controlled to work allon formalin-fixed paraffin-embedded as well as acetone fixed cryostat tissue sections. No further titration is required. We suggest an incubation period of 30-60 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation conditions should be determined by the user. Enzymatic treatment with proteolytic enzyme may be required with certain tissues.

Type: Primary
Antigen: IGG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR