Anti-IgG Rat Monoclonal Antibody [clone: COCKTAIL]

Supplier: GeneTex

GTX76129
89367-966EA 785.42 CAD
89367-966
Anti-IgG Rat Monoclonal Antibody [clone: COCKTAIL]
Antibodies
Rat anti-Mouse IgG Monoclonal antibody [COCKTAIL]

Recommended Starting Dilutions: ELISA: Max Dilution 1.25 ug/ml Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: IGG
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR