Anti-IgG Rat Monoclonal Antibody [clone: COCKTAIL] (Biotin)

Supplier: GeneTex
GTX76130
89367-968EA 914.5 CAD
89367-968
Anti-IgG Rat Monoclonal Antibody [clone: COCKTAIL] (Biotin)
Antibodies
Biotin-conjugated Rat anti-Mouse IgG Monoclonal antibody [COCKTAIL]

Recommended Starting Dilutions: ELISA: Max Dilution 1.0 ug/ml Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy chain
Host: Rat
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR