Anti-IgM Mouse Monoclonal Antibody [clone: CM7]

Supplier: GeneTex

GTX75506
89367-884EA 1039.56 CAD
89367-884
Anti-IgM Mouse Monoclonal Antibody [clone: CM7]
Antibodies
Mouse monoclonal antibody [CM7] to IgM

Recommended Starting Dilutions: Flow Cytometry: 1/10 - 1/50 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Secondary
Antigen: IgM
Clonality: Monoclonal
Clone: CM7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR