Anti-IL1A Mouse Monoclonal Antibody

GTX10747
89354-964EA 427.1 CAD
89354-964
Anti-IL1A Mouse Monoclonal Antibody
Interleukin 1 (IL1), originally known as lymphocyte activating factor (LAF), activates T cells and lymphocytes, which then proliferate and secrete interleukin 2. IL1 is primarily released from stimulated macrophages and monocytes, but also is released from several other cell types and is thought to play a key role in inflammatory and immune responses. Other synonyms for IL1 include: endogenous pyrogen (EP), mitogenic protein (MP), helper peak 1 (HP1), T cell replacing factor III (TRF III or TRFH), B cell activating factor (BAF) and B cell differentiation factor (BDF). The two closely related agents, interleukin 1alpha (IL1 alpha) and interleukin1 beta (IL1 beta) bind to the same cell surface receptor, elicit nearly identical biological responses and share 25% homology in their amino acid sequence. The two closely related agents, interleukin 1 alpha (IL1 alpha) and interleukin 1 beta (IL1 beta) bind to the same cell surface receptor, elicit nearly identical biological responses and share 25% homology in their amino acid sequence.

Type: Primary
Antigen: IL1A
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR