Anti-IL1B Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX59836
89363-744EA 870.66 CAD
89363-744
Anti-IL1B Rabbit Polyclonal Antibody
Antibodies
IL-1beta is a proinflammatory cytokine produced in a variety of cells including monocytes, tissue macrophages, keratinocytes and other epithelial cells. Both IL-1alpha and IL-1beta binds to the same receptor and has similar if not identical biological properties. These cytokines have a broad range of activities including, stimulation of thymocyte proliferation, by inducing IL-2 release, B-cell maturation and proliferation, mitogenic FGF-like activity and the ability to stimulate the release of prostaglandin and collagenase from synovial cells. However, whereas IL-1beta is a secreted cytokine, IL-1alpha is predominantly a cell-associated cytokine. Recombinant murine IL-1beta is a 17.5 kDa protein containing 153 amino acid residues.

Type: Primary
Antigen: IL1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR