Anti-IL2 Mouse Monoclonal Antibody [clone: 14-0]

Supplier: GeneTex
GTX10089
89350-624EA 1147.57 CAD
89350-624
Anti-IL2 Mouse Monoclonal Antibody [clone: 14-0]
Antibodies
Mouse monoclonal [14 - 0] to IL2

ELISA: Use at an assay dependent dilution. Suitable for sandwich ELISA of IL2 in the following combination: capture: GTX10090 / conjugate: GTX10089. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IL2
Clonality: Monoclonal
Clone: 14-0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR