Anti-IL2RG Rabbit Polyclonal Antibody

GTX102717
89349-702EA 272.33 CAD
89349-702
Anti-IL2RG Rabbit Polyclonal Antibody
The interleukin 2 (IL2) receptor gamma chain (IL2RG), an important signalling component of many interleukin receptors (IL2,IL4,IL7,IL9, and IL15), is thus referred to as the common gamma chain. Mutations in this X-chromosome-linked gene cause X-linked severe combined immunodeficiency (XSCID). [provided by RefSeq]

Type: Primary
Antigen: IL2RG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR