Anti-IL3 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX29620
89367-640EA 837.04 CAD
89367-640
Anti-IL3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to IL3

Neutralization: 1 - 10 ?g/ml. WB: 0.5 - 1 ?g/ml. However, the optimal conditions should be determined individually.

Type: Primary
Antigen: IL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR