Anti-IL4 Mouse Monoclonal Antibody [clone: 14916]

Supplier: GeneTex
GTX10106
89350-628EA 533.93 CAD
89350-628
Anti-IL4 Mouse Monoclonal Antibody [clone: 14916]
Antibodies
Mouse monoclonal [11 - 40] to IL4

ELISA: Use at an assay dependent dilution. Can be used in a ELISA pair with GTX10107 (conjugate). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IL4
Clonality: Monoclonal
Clone: 14916
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR