Anti-IL6R Rabbit Polyclonal Antibody

Supplier: Genetex
GTX103966
89350-174EA 1813 CAD
89350-174
Anti-IL6R Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to IL6 Receptor (interleukin 6 receptor)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: IL6R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR