Anti-ILK Rabbit Monoclonal Antibody

GTX62096
89364-316EA 556.26 CAD
89364-316
Anti-ILK Rabbit Monoclonal Antibody
Integrin-linked kinase (ILK) is a key cytoplasmic plaque protein involved in the mediation of integrin functions. It plays a key role as a regulator during cell survival, cell proliferation, cell migration, and cell to cell adhesion . ILK links integrins to the actin cytoskeleton, allowing transduction of signals through integrins to the extracellular matrix. Also, ILK has been linked to the regulation of smooth contraction and cell motility in non-muscle cells through phosphorylation of myosin . Irregular activity level of ILK has been linked to expression of certain tumor. ILK overexpression caused by inactivating mutation in tumor suppressors (e.g. PTEN and APC) lead to tumor formation in transgenic mouse. Up-regulated ILK expression and activity are seen in Ewings scaromas, colorectal cancers, and late staged prostate cancers .

Type: Primary
Antigen: ILK
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR