Anti-ITGB1 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX25189
89361-590EA 719.9 CAD
89361-590
Anti-ITGB1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Integrin beta 1 ( phospho T788 + T789 )

WB: Use at a concentration of 0.1 - 1.0 ?g/ml. Predicted molecular weight: 130 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ITGB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR