Anti-IFNA1 Mouse Monoclonal Antibody [clone: AE3]

Supplier: GeneTex

GTX10077
89350-620EA 439.98 CAD
89350-620
Anti-IFNA1 Mouse Monoclonal Antibody [clone: AE3]
Antibodies
Mouse monoclonal [AE3] to Interferon alpha

ELISA: Use at an assay dependent dilution. IA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 23 kDa. Can be used as an ELISA pair wih GTX10078 (conjugate). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IFNA1
Clonality: Monoclonal
Clone: AE3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR