Anti-Invertase Goat Polyclonal Antibody

Supplier: GeneTex
GTX40597
89363-256EA 1369.12 CAD
89363-256
Anti-Invertase Goat Polyclonal Antibody
Antibodies
Invertase hydrolyses the terminal non-reducing beta-D-fructofuranoside residues in beta-D-fructofuranosides. It is the enzyme that bees use to convert nectar into honey. Industrial / confectionary applications use yeast invertase to split sucrose into fructose and glucose and also to improve the shelf life of food products.

Type: Primary
Antigen: Invertase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR