Anti-IRF3 Rabbit Monoclonal Antibody

GTX62132
89364-324EA 584.3 CAD
89364-324
Anti-IRF3 Rabbit Monoclonal Antibody
A family of interferon (IFN) regulatory factors (IRFs) have been shown to play a role in transcription of IFN genes as well as IFN-stimulated genes. The IRF-3 gene encodes a 50-kDa protein that binds specifically to the IFN-stimulated response element (ISRE) . IRF-3 is activated through phosphorylation by the kinases IKK epsilon and/or TBK1. Phosphorylation results in IRF-3 dimerization and removal of an autoinhibitory structure to allow interaction with the coactivators CBP/p300. Phosphorylation at Serine 396 through Serine 405 alleviates autoinhibition to allow interaction with CREB-binding protein and facilitates phosphorylation at Serine 385 or Serine 386 . Serine 386 is suggested to be the target of the IRF-3 kinase and critical determinant for the activation of IRF-3 .

Type: Primary
Antigen: IRF3 (Phospho SER386)
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR