Anti-IRF5 Goat Polyclonal Antibody

Supplier: GeneTex
GTX22932
89361-328EA 622.46 CAD
89361-328
Anti-IRF5 Goat Polyclonal Antibody
Goat polyclonal to IRF5

Western Blot - Approx 55-60kDa band seen in A549 lysate (predicted MW 58 kDa). Recommended for use at 1.0-3.0ug/ml.

Type: Primary
Antigen: IRF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR