Anti-ITK Mouse Monoclonal Antibody [clone: 2F12]

Supplier: GeneTex

GTX23302
89367-068EA 818.91 CAD
89367-068
Anti-ITK Mouse Monoclonal Antibody [clone: 2F12]
Antibodies
Mouse monoclonal [2F12] to ITK

Immunoprecipitation (with protein G): use at a concentration of 2?g / mg of lysate. Western Blotting: use at a concentration of 1 - 2?g / ml for 2hrs at RT. Not tested in other applications. Optimal dilutions / concentrations should be determined by the end user. By Western blot, this antibody detects a band of ~ 72 kDa, which corresponds to the predicted molecular weight of ITK.

Type: Primary
Antigen: ITK
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR