Anti-ITSN1 Chicken Polyclonal Antibody

GTX82134
89368-054EA 666.36 CAD
89368-054
Anti-ITSN1 Chicken Polyclonal Antibody
Adapter protein that may provide indirect link between the endocytic membrane traffic and the actin assembly machinery. May regulate the formation of clathrin-coated vesicles. Isoform 1 could be involved in brain-specific synaptic vesicle recycling.

Type: Primary
Antigen: ITSN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR