Anti-JAM2 Rat Monoclonal Antibody

Supplier: Genetex
GTX75970
89365-946EA 513.52 CAD
89365-946
Anti-JAM2 Rat Monoclonal Antibody
Junctional Adhesion Molecule C (JAMC) belongs to the immunoglobulin superfamily is a 45kD cell adhesion molecule found at intercellular junctions of endothelial cells. It plays an important role in tight junctions where it is involved in cell-to-cell adhesion through homophilic interaction. It co distributes with other tight junction componenets such as ZO-1, 7H6 antigen, cingulin and occludin.

Type: Primary
Antigen: JAM2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR