Anti-JIK / TAOK3 Goat Polyclonal Antibody

Supplier: Genetex
GTX89693
89367-004EA 919.07 CAD
89367-004
Anti-JIK / TAOK3 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to JIK / TAOK3

Approx 100kDa band observed in human Liver lysate (calculated MW of 105kDa according to NP_057365). Recommended concentration: 0.3-1.0 µg/ml. In paraffin embedded Human Prostate shows staining of cytoplasm of secretory cells in the glands. Recommended concentration, 2-3µg/ml.

Type: Primary
Antigen: JIK / TAOK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR