Anti-MAPK9 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX107817
89355-156EA 595.28 CAD
89355-156
Anti-MAPK9 Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to JNK2 (mitogen-activated protein kinase 9)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MAPK9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR