Anti-CDH6 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX74069
89367-750EA 933.05 CAD
89367-750
Anti-CDH6 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to CXCL1

Recommended Starting Dilutions: ELISA: 0.5 - 2.0 ug/ml Western Blotting: 0.1 - 0.2 ug/ml Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CDH6
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR