Anti-KI67 Rabbit Monoclonal Antibody

GTX62301
89364-388EA 556.26 CAD
89364-388
Anti-KI67 Rabbit Monoclonal Antibody
Ki-67 antigen is a nuclear antigen specifically associated with cell proliferation. Ki-67 is expressed in all proliferating cells which are in the active phases of the cell cycle (late G1, S, G2, and mitosis), but absent in resting cells. In G1, it is predominately localized in the perinucleolar region and also found in nuclear matrix in the later phases . Ki-67 is thought to be involved in the maintaining of cell proliferation, however, its exact mechanism for function is unknown. Due to the fact that Ki-67 can be used to determine the growth fraction (the proportion of the cells committed to the cell cycle), it is consider an important marker in human tumor diagnostics and prognostics .

Type: Primary
Antigen: KI67
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR