Anti-KIDINS220 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX70435
89364-982EA 580.76 CAD
89364-982
Anti-KIDINS220 Rabbit Polyclonal Antibody
Antibodies
A novel protein kinase D-interacting substrate of 220 kDa (Kidins220) is the first identified Protein kinase D (PKD), a serine/threonine kinase. Kidins220 is an integral membrane protein that is expressed in brain and neuroendocrine cells. Kidins220 is phsphorylated by PKD at serine 919 in vivo.

Type: Primary
Antigen: KIDINS220
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR