Anti-KIF14 Rabbit Polyclonal Antibody

GTX23746
89352-050EA 479.79 CAD
89352-050
Anti-KIF14 Rabbit Polyclonal Antibody
KIF14 is a member of the kinesin superfamily of microtubule-associated motors (see MIM 148760) that play important roles in intracellular transport and cell division (Nakagawa et al., 1997 [PubMed 9275178]).[supplied by OMIM]

Type: Primary
Antigen: KIF14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR