Anti-KLF2 Rabbit Polyclonal Antibody

GTX104834
89353-972EA 258.54 CAD
89353-972
Anti-KLF2 Rabbit Polyclonal Antibody
Binds to the CACCC box in the beta-globin gene promoter and activates transcription.

Type: Primary
Antigen: KLF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR