Anti-LF Rabbit Monoclonal Antibody

GTX62696
89364-570EA 556.26 CAD
89364-570
Anti-LF Rabbit Monoclonal Antibody
Lactoferrin is a member of the transferrin family that is found in secondary granules of neutrophils. The protein is a major iron-binding protein in milk and body secretions with antimicrobial activity, making it an important component of the non-specific immune system. Lactoferrin demonstrates a broad spectrum of properties: regulation of iron homeostasis, host defense against a broad range of microbial infections, anti-inflammatory activity, regulation of cellular growth and differentiation, and protection against cancer development and metastasis .

Type: Primary
Antigen: LF
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR