Anti-LMNA Rabbit Monoclonal Antibody

GTX62501
89364-474EA 556.26 CAD
89364-474
Anti-LMNA Rabbit Monoclonal Antibody
The nuclear lamina consists of a two-dimensional matrix of proteins located next to the inner nuclear membrane. The lamin family of proteins makes up the matrix and is highly conserved in evolution. During mitosis, the lamina matrix is reversibly disassembled as the lamin proteins are phosphorylated. Lamin proteins are thought to be involved in nuclear stability, chromatin structure and gene expression. Vertebrate lamins consist of two types, A and B . By alternate splicing, lamin A/C produces the following three isoforms: isoform A (74 kDa), isoform C (65 kDa), and isoform ADelta10 (70 kDa) . Mutations in lamin A/C lead to several diseases: Emery-Dreifuss muscular dystrophy, familial partial lipodystrophy, limb girdle muscular dystrophy, dilated cardiomyopathy, Charcot-Marie-Tooth disease, and Hutchinson-Gilford progeria syndrome .

Type: Primary
Antigen: LMNA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR