Anti-LCK Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX24900
89361-574EA 666.57 CAD
89361-574
Anti-LCK Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Lck ( phospho Y192 )

WB: Use at a concentration of 0.1 - 1.0 µg/ml. Predicted molecular weight: 60 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: LCK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR