Anti-LCK Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX24901
89361-576EA 920.44 CAD
89361-576
Anti-LCK Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Lck ( phospho Y505 )

WB: Use at a concentration of 0.1 - 1.0 µg/ml. Predicted molecular weight: 60 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: LCK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR