Anti-LCK Mouse Monoclonal Antibody [clone: LCK-01]

Supplier: GeneTex

GTX28075
89362-718EA 622.58 CAD
89362-718
Anti-LCK Mouse Monoclonal Antibody [clone: LCK-01]
Antibodies
Mouse monoclonal [LCK - 01] to Lck

FACS: Use at an assay dependant dilution. ICC: Use at an assay dependant dilution. IP: Use at an assay dependant concentration. WB: Use at an assay dependant concentration. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: LCK
Clonality: Monoclonal
Clone: LCK-01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR