Anti-LEP Mouse Monoclonal Antibody

GTX26055
89352-154EA 374.88 CAD
89352-154
Anti-LEP Mouse Monoclonal Antibody
Leptin, the product of the ob (obese) gene, is a 16 kDa protein consisting of 146 amino acid residues. Leptin is produced in the adipose tissue, and is considered to play an important role in appetite control, fat metabolism and regulation of body weight. It targets the central nervous system, particularly hypothalamus, affecting food intake. Leptin levels are high in most obese individuals. Studies have shown that it may also influence reproductive function.

Type: Primary
Antigen: LEP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR