Anti-L-MYC Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX48739
89363-682EA 937.18 CAD
89363-682
Anti-L-MYC Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to l-Myc

IHC-P: Use at a concentration of 4 - 8 ?g/ml. WB: Use at a concentration of 2.5 ?g/ml. Predicted molecular weight: 40/25 kDa.

Type: Primary
Antigen: L-MYC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR