Anti-LRP6 Rabbit Monoclonal Antibody

GTX62901
89364-604EA 556.26 CAD
89364-604
Anti-LRP6 Rabbit Monoclonal Antibody
LRP6 (low-density lipoprotein receptor-related protein 6) is a single span transmembrane Wnt receptor that interacts with Axin to promote the Wnt-dependent accumulation of beta-catenin. Cells treated with Wnt quickly induce plasma membrane-associated LRP6 aggregates. The aggregates are phosphorylated, and then promote Axin recruitment and beta-catenin stabilization. LRP6 is internalized with caveolin and components of this endocytic pathway are required not only for Wnt-3a-induced internalization of LRP6 but also for accumulation of beta-catenin (1-2).

Type: Primary
Antigen: LRP6
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR