Anti-LSM1 Chicken Polyclonal Antibody

Supplier: GeneTex
GTX57463
89367-742EA 978.73 CAD
89367-742
Anti-LSM1 Chicken Polyclonal Antibody
Antibodies
Chicken polyclonal to LSM1

For WB: Use at an assay dependent dilution. Predicted molecular weight: 15 kDa. We recommend using about 2.5 ug of positive control protein in the colorimetric method, and about 10x less for ECL detection. Not yet tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: LSM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR