Anti-LXN Chicken Polyclonal Antibody

GTX14057
89367-254EA 642.96 CAD
89367-254
Anti-LXN Chicken Polyclonal Antibody
Latexin, a protein possessing inhibitory activity against rat carboxypeptidase A1 (CPA1) and CPA2, is expressed in a neuronal subset in the cerebral cortex and cells in other neural and non-neural tissues of rat. Although latexin also inhibits mast-cell CPA (MCCPA), the expression of latexin in rat mast cells has not previously been confirmed.

For ICC: Use at a dilution of 1:100 - 1:300. For IHC: Use at an assay dependent dilution. Predicted molecular weight: 25 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: LXN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR