Anti-CDH15 Mouse Monoclonal Antibody [clone: 21G4]

GTX44915
89363-560EA 754.68 CAD
89363-560
Anti-CDH15 Mouse Monoclonal Antibody [clone: 21G4]
Mouse monoclonal [21G4] to M Cadherin

ELISA: Use at a concentration of 0.05 ?g/ml. IF: Use at a concentration of 5 ?g/ml. WB: Use at a concentration of 0.5 - 1 ?g/ml. (1ug/ml for AP/BCIP/NBT (MTT) detection; 0.5ug/ml for HRP/ECL detection). Predicted molecular weight: 90 kDa. Not yet tested in other applications.Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CDH15
Clonality: Monoclonal
Clone: 21G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR